Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GR-M

95032301, human skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

GR-M, 95032301

biological source

human skin

growth mode

Adherent

karyotype

Modal no. 60-64

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian melanoma

Cell Line Description

GR-M was obtained from the thorax of a 36 year old male with skin metastases having received no treatment at the time of the biopsy. The cells express HLA class II and the melanoma associated antigen P97. The expression of epithelial cell kinase (ECK), a receptor protein tyrosine kinase, was also detected.

Application

Carcinogenesis studies.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Heated Inactivated Foetal Bovine Serum (HI-FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service