Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AFRC 84C6

Reactivity: Anti-Haemagglutinin, influenza, 94120830

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

biological source

Horse x Mouse/Horse Hybridoma

ImmunoDon

Horse sub-mandibular lymph node

Myeloma

AFRC 17D1

Reactivity

Anti-Haemagglutinin, influenza

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Storage temp.

−196°C

Cell Line Origin

Horse sub-mandibular lymph node x AFRC 17D1

Cell Line Description

The heterohybridoma AFRC 84C6 is one of 8 hybridoma cell lines obtained using lymphocytes from ponies immunised with influenza A equine 2 virus (isolateA/Equine/Newmarket/79 (H3N8)). For the establishment of AFRC 84C6 horse cells from the sub-mandibular lymph node have been fused with the non-Ig-secreting heterohybridoma AFRC 17D1 (Sigma Catalogue number. 94120825).

Immunogen

Influenza A equine 2 virus

Application

Haemagglutinin detection
The isotype of the monoclonal antibody produced by this cell line is horse IgG.

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 5% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service