Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COLO 775

94072236, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

COLO 775, 94072236

biological source

human blood

growth mode

Suspension

karyotype

Modal no. 65, XXY

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Storage temp.

−196°C

Cell Line Origin

Human leukaemia

Cell Line Description

COLO 775 was established in 1990 from the peripheral blood cells of a 57 year old female with chronic myelogenous leukaemia. The cells show a complex karyotype with some characteristics of myelogenous leukaemia.

Application

Carcinogenesis studies, chromosome analysis

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service