GRL101 (KC7)
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
hamster ovary
packaging
tube of 5 μg 94071259-DNA-5UG
pkg of vial of cells 94071259-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
GRL101 - putative Lymnaea receptor
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Hamster Chinese ovary transfected with Lymnaea receptor GRL101
Cell Line Description
Cell line containing the receptor protein GRL101. The cDNA from the CNS of the mollusc Lymnaea stagnalis encoding a putative receptor has been isolated and analysed. The first repeat of DNA displays a high sequence similarity to the extracellular binding domains of the low density protein receptor, which binds and internalises cholesterol containing apolipoproteins. The MRNI receptor is predominantly expressed in a small number of neurons within the CNS and to a lesser extent in the heart.
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2, 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service