Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GRL101 (MIX)

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

hamster ovary

packaging

tube of 5 μg 94071258-DNA-5UG
pkg of vial of cells 94071258-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

GRL101 - putative Lymnaea receptor

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese ovary transfected with Lymnaea receptor GRL101

Cell Line Description

Cell line containing the receptor protein GRL101. The cDNA from the CNS of the mollusc Lymnaea stagnalis encoding a putative receptor has been isolated and analysed. The first repeat of DNA displays a high sequence similarity to the extracellular binding of the low density protein receptor, which binds and internalises cholesterol containing apolipoproteins. The MRNI receptor is predominantly expressed in a small number of neurons within the CNS and to a lesser extent in the heart. Contains a mixed culture of approximately 20 transfected clones.

Application

Studies in the newly characterised members of a putative class of G protein - linked receptors that directly transduce signals carried by large extracellular (lipo)protein (complexes) into neural events.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2 - 4 x 10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2, 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service