Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3/25

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 94050507-DNA-5UG
pkg of vial of cells 94050507-1VL

ImmunoDon

Mouse spleen

Myeloma

S194/5.XXO.BU.1

Reactivity

Anti-Haematopoietic Cell, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse S194/5.XXO.BU.1 x Mouse spleen cells

Cell Line Description

The antibody is directed against an acidic glycoprotein (dimer of 2x110kDa monomers joined by disulphide bonds) present on immature and proliferating human haematopoietic cells but not circulating peripheral blood cells, or other non haematopoietic cells.

Immunogen

K-562 cells

Culture Medium

DMEM + 2mM glutamine + 0.02mM 2-mercaptoethanol (2ME) + 10% Horse Serum.

Subculture Routine

Maintain cultures between 100,000 - 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service