OVNIS
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
ovine thyroid
packaging
tube of 5 μg 93122324-DNA-5UG
pkg of vial of cells 93122324-1VL
morphology
Epithelial
products
Thyroglobulin, plasminogen activators (t-PA and u-PA)
receptors
Thyrotropin, insulin, hydrocortisone, Insulin Growth Factor (IGF)-I
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Ovine thyroid epithelial
Cell Line Description
Isolated from ovine thyroid and adapted to grow in low serum (0.1%) media.
Application
Regulation of thyroid cell metabolism
Culture Medium
Ham′s F12 + 121mg/l CaCl2(2H20) + 10μg/ml Insulin + 10nM Hydrocortisone + 5μg/ml Transferrin + 10ng/ml Glycyl-L-histydyl-L-lysine acetate + 10ng/ml somatostatin + 10mU/ml Thyroid Stimulating Hormone (TSH) + 2mM Glutamine + 0.1%Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 using 0.05% trypsin/0.025% EDTA in Puck F saline; 2.5% CO2; 37°C.
Other Notes
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service