VPM 44
Reactivity: Anti-MHC class II DQβ chain, ovine, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93113033
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 93113033-DNA-5UG
pkg of vial of cells 93113033-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
NS0
Reactivity
Anti-MHC class II DQβ chain, ovine
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS0 x Mouse BALB/c spleen
Cell Line Description
The antibody is directed against MHC class II molecules on cell surface and on immunoblots. It also reacts with polymorphic determinant and is therefore subgroup specific.
Immunogen
Ovine MHC class II
Application
Identifying and characterising sheep MHC class II molecules
Culture Medium
RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 1-2x100,000 cells/ml; 5% CO2; 37°C. Saturation density reached at 3-9x100,000 cells/ml. (takes 3-4 days to reach 5x100,000)
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service