VPM 38
93113029, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-MHC class II α-chain, ovine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 93113029-DNA-5UG
pkg of vial of cells 93113029-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
NS0
Reactivity
Anti-MHC class II α-chain, ovine
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS0 x Mouse BALB/c spleen
Cell Line Description
The antibody cross-reacts with bovine and human MHC class II. It reacts with MHC class II molecules on cell surfaces and on immunoblots. It reacts very weakly with ovine DQ L cell transfectants by immunofluorescence, but does not react with DQ blots and therefore is classified as strongly DR α specific, but weakly cross-reactive with DQ α.
Immunogen
Ovine MHC class II
Application
Quantification of DQ and DR expression on T cell subsets in adult and foetal peripheral blood, afferent lymph, lymph node and efferent lymph.
Culture Medium
RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 1-2x100,000 cells/ml; 5% CO2; 37°C. Saturation density is reached at 3-9x100,000 cells/ml.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service