The medium for each cell line is listed on the product page. It is specific to each cell line. If not visible on the Sigma-Aldrich website product page, the HPA website also contains the same information (www.hpacultures.org.uk). The product number for the cell line is the same on either site.
A2780cis Cell Line human
93112517, human ovary, Epithelial
Synonym(s):
A2780cis Cells
About This Item
Recommended Products
Product Name
A2780cis Cell Line human, ovarian carcinoma, 93112517
biological source
human ovary
growth mode
Adherent
karyotype
Modal no. 46
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Application
DNA Profile
CSF1PO: 10,11
D13S317: 13
D16S539: 11,13
D5S818: 11
D7S820: 10
THO1: 6
TPOX: 8,10
vWA: 15,16
Culture Medium
Subculture Routine
Other Notes
Disclaimer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What medium should I use when I thaw the Cell Line cells?
1 answer-
Helpful?
-
-
Are my Cell Line cells mycoplasma tested?
1 answer-
ECACC routinely tests all manufactured cell banks for mycoplasma.
Helpful?
-
-
How do I handle Cell Line frozen cells upon arrival?
1 answer-
Upon receipt, frozen ampules should be transferred directly to vapor phase liquid nitrogen without delay (-135°C) or liquid phase liquid nitrogen if vapor phase is not available. DO NOT use a -80°C freezer as an alternative; this will result in loss of viability.
Helpful?
-
-
Are the Cell Line cells tested for viral pathogens?
1 answer-
HPA Cultures does not perform any viral testing on the cell lines.
Helpful?
-
-
How many Cell Line cells are in the vial?
1 answer-
Each vial of cells contains 2-3 x 106 cells in 1 ml of freezing media. This is in a 1.8 mL ampule. For exceptionally large cells, counts may decrease. Suspension cells, generally smaller cells, may contain as many as 4-5 x 106 cells/vial to assure optimal viability upon thaw.
Helpful?
-
-
What passage are my Cell Line cells?
1 answer-
If the passage of the cells is known, it is listed on the product page at the HPA website (www.hpacultures.org.uk).
Helpful?
-
-
Why is Vapor Phase Liquid Nitrogen preferred for storage of Cell Line cells?
1 answer-
If ampules are immersed into liquid phase of liquid nitrogen, it increases the risk of the liquid seeping into the vial. This could lead to problems of cross-contamination and increased risk of the ampule exploding when thawed.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service