Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS/2

93091302, Reactivity: Anti-VLA-4 integrin-like cell adhesion molecule, murine, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Rat Hybridoma

packaging

tube of 5 μg 93091302-DNA-5UG
pkg of vial of cells 93091302-1VL

ImmunoDon

Fischer Rat spleen

Myeloma

Sp2/0

Reactivity

Anti-VLA-4 integrin-like cell adhesion molecule, murine

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Rat Fischer spleen

Cell Line Description

The antibody is directed against the α-4 chain of the murine VLA-4 integrin-like cell adhesion molecule which is present on blood cells. The antibody will react with human integrins.

Immunogen

P815 murine mastocytoma cells

Application

The isotype of the monoclonal antibody produced by this cell line is rat IgG2b.

Culture Medium

RPMI 1640 + 2mM glutamine + 0.05mM 2-mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service