Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AFRC MAC 158

93090998, Reactivity: Anti-Cytokinin, plant, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Rat Hybridoma

packaging

tube of 5 μg 93090998-DNA-5UG
pkg of vial of cells 93090998-1VL

ImmunoDon

Rat F344 spleen

Myeloma

NS0

Reactivity

Anti-Cytokinin, plant

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS0 x Rat F344 spleen

Cell Line Description

The antibody is directed against the plant hormone isopentenyladenosine.

Immunogen

Isopentenyladeonsine cytokinin conjugate and BSA

Application

Detection of 2iPA in plant tissue
The isotype of the monoclonal antibody produced by this cell line is rat IgM.

Culture Medium

DMEM + 2mM glutamine + 20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service