Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FW 11.10.3

Reactivity: Anti-CD44 v4, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93070776

Sign Into View Organizational & Contract Pricing

Select a Size

93070776-DNA-5UG
$402.00
1 VIAL
$981.00

$402.00


Usually ships in 1 week. (Orders outside of Europe & Scandinavian countries, allow an additional week for delivery


Select a Size

Change View
93070776-DNA-5UG
$402.00
1 VIAL
$981.00

About This Item

UNSPSC Code:
41106514

$402.00


Usually ships in 1 week. (Orders outside of Europe & Scandinavian countries, allow an additional week for delivery

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 93070776-DNA-5UG
pkg of vial of cells 93070776-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-CD44 v4, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c x Mouse Sp2/0 myeloma

Cell Line Description

The antibody is directed against the variant region of human CD44 encoded by exon 4. Staining binding must be performed at room temperature.

Immunogen

Fusion protein of human CD44

Application

Immunohistology and FACS

Culture Medium

IDMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service