Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IC-07

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-Galactocerebroside, chicken, 93062301

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 93062301-DNA-5UG
pkg of vial of cells 93062301-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3-NS1/1-Ag4.1

Reactivity

Anti-Galactocerebroside, chicken

technique(s)

cell culture | mammalian: suitable

isotype

IgG3

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3-NS1/1-Ag4.1 x Mouse BALB/c spleen

Cell Line Description

The antibody is reported to be directed against galactocerebroside present on oligodendrocytes and Schwann cells. The antibody is also reported to cross-react with related molecules in rodents and fish. Originally this antibody was reported to have an IgG1 subclass but testing at ECACC has shown it to be IgG3. However, it has not been possible to validate the antibody.

Immunogen

Chicken synaptosomes

Application

Immunohistology

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service