Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.6.5a114K

93062218, Reactivity: Anti-Madin-Darby canine kidney cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0 Ag-14

Reactivity

Anti-Madin-Darby canine kidney cells

technique(s)

cell culture | mammalian: suitable

isotype

IgG1, kappa

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 Ag-14 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against a 144kDa protein present predominantly on the apical, but also on the basolateral surfaces of Madin-Darby canine kidney cells of the MDCK cell line.

Immunogen

Basolateral membrane fraction from dog intestine

Application

Study of cell surface polarity development

Culture Medium

EMEM(EBSS) + 2mM glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service