Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COLO 720 L

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human blood

packaging

tube of 5 μg 93052623-DNA-5UG
pkg of vial of cells 93052623-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human normal B lymphocytes

Cell Line Description

Normal B lymphoid cell line derived from the peripheral blood of a 74 year old Chinese female (the same patient as COLO 720E) with ovarian cancer. These B-lymphoid cells grow as characteristic clumps of cells in the media. Often the peripheral cells of the clumps have their spike-like uropods extending into the media.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 8,11
D16S539: 11
D5S818: 10,11
D7S820: 10
THO1: 9
TPOX: 11
vWA: 14

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service