Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COLO 206F

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 93052620, human colon, Predominantly round, refractile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human colon

packaging

tube of 5 μg 93052620-DNA-5UG
pkg of vial of cells 93052620-1VL

growth mode

Semi-adherent

karyotype

Not specified

morphology

Predominantly round, refractile

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon carcinoma

Cell Line Description

COLO 206F, COLO 201 (ECACC Catalogue 87091201) and COLO 205 (ECACC Catalogue 87061208) were all established from the ascites fluid obtained from a 70-year-old male patient with adenocarcinoma of the colon (Dukes Classification Grade D). The cell line can be used for immunological studies in cytotoxicity assays and immunoglobin characterisation. Please note this cell line was mycoplasma eradicated at ECACC.

Culture Medium

RPM11640, 10% Foetal Bovine Serum (FBS) + 2mM Glutamine.

Subculture Routine

Maintain cultures between 3-9x105 cells/ml 5% CO2, 37°C. Any attached cells can be dislodged with trypsin/EDTA.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service