Z2
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-IgG2a, mouse, 93022522
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 93022522-DNA-5UG
pkg of vial of cells 93022522-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0
Reactivity
Anti-IgG2a, mouse
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Sp2/0 x Mouse BALB/c spleen
Cell Line Description
The immunisation protocol involved repeated intranasal infection with a single i.v. injection before fusion. Produces IgG2b antibody which binds murine IgG2a of "a" and "b" allotype, Fc region. There is no cross-reactivity with A/PR/8/34 influenza virus, DNP, NP, DNA or cytoskeletal proteins.
Immunogen
A/PR/8/34 mouse pathogenic human influenza virus
Application
Differential detection of complexed IgG2a in the presence of native IgG2a, allotype detection or quantitation.
Culture Medium
RPMI 1640 or DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS) (can be adapted to 1% Foetal Bovine Serum (FBS))
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service