Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0131

from human blood, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93021104

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human blood

packaging

tube of 5 μg 93021104-DNA-5UG
pkg of vial of cells 93021104-1VL

HRC Panel Number

3

ethnicity

UK Caucasian

gender

female

growth mode

Suspension

karyotype

Not Specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

UK Caucasian Female

Cell Line Description

A B-lymphoblastoid cell line derived from a randomly selected Caucasian blood donor whose parents and grandparents were born in the UK or Ireland. The cell line was generated by Epstein-Barr Virus (EBV) transformation of peripheral blood lymphocytes.

This cell line is part of the European Collection of Cell Cultures (ECACC) Human Random Control (HRC) collection. The HRC collection consists of more than 700 B-lymphoblastoid cell lines derived from randomly selected Caucasian blood donors whose parents and grandparents were born in the UK or Ireland. The HRC collection can be used as a renewable control population for genetic research.

application

As part of the Human Random Control collection this cell line can be used as a renewable source of genomic DNA reference material for a wide range of genetic research applications.

Culture Medium

RPMI 1640 (R0883) + 2 mM L-Glutamine (G7513) + 20% FBS / FCS (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.

Subculture Routine

Maintain cultures between 3 x 105 - 2 x 106 cells/ml; 5% CO2, 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service