Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HyGPD-YK-1-1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 92120704-DNA-5UG
pkg of vial of cells 92120704-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-Muscle Glyceraldehyde-3-phosphate Dehydrogenase, rabbit

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P363Ag8.653 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against rabbit muscle glyceraldehyde-3-phosphate dehydrogenase. This line is a World Reference cell line capable of growing in serum-containing and serum-free media (supplemented with insulin, transferrin, ethanolamine and selenium). The ampoules supplied are from an 180 ampoule distribution stock prepared at the Riken Cell Bank in Japan.

Immunogen

Rabbit glyceraldehyde-3-phosphate dehydrogenase

Application

Comparison of newley developed bioreactions, media, media components etc.

Culture Medium

E-RD7 + 2mM glutamine + 5% Foetal Bovine Serum (FBS) (Kyokuto) or serum-free RPMI 1640

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service