Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NH17

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human blood

packaging

tube of 5 μg 92111101-DNA-5UG
pkg of vial of cells 92111101-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

High expresser of surface HTA1

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T cell lymphoblastic leukaemia

Cell Line Description

Subclone of MOLT-4 (Sigma Catalogue number. 85011413) and is a high expresser of the cortical thymocyte differentiation antigen HTA1 and can be used for the production of CD1DmRNA.

Application

Production of CD1DmRNA

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells should not remain in culture for long periods otherwise expression of CD1 is lost.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service