Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AFRC MAC 22

92102336, Reactivity: Anti-LDL, rabbit, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Rat x Rat Hybridoma

packaging

tube of 5 μg 92102336-DNA-5UG
pkg of vial of cells 92102336-1VL

ImmunoDon

LOU rat spleen

Myeloma

Y3

Reactivity

Anti-LDL, rabbit

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Y3 x Rat LOU spleen

Cell Line Description

The antibody is directed against rabbit low density lipoproteins (LDL), and shows 11-34% reactivity with murine LDL.

Immunogen

Rabbit low density lipoproteins

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse spleen feeder layers upon resuscitation

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service