Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RENT ts3.1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

rat endometrium

packaging

tube of 5 μg 92092516-DNA-5UG
pkg of vial of cells 92092516-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Cuboidal

products

Not specified

receptors

Glucocorticoid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Sprague-Dawley, primary endometrial, SV40 T antigen transformed

Cell Line Description

Derived from the endometrium tissue from the uterus of a 21-28 day-old Sprague-Dawley rat, transformed with a ts mutant of SV40 T antigen (tsA58). Cells express vimentin, CSF-1 (at 39°C), desmin, collagen type III and connexin 43.

Culture Medium

DMEM + 20μg/l Gly-His-Lys (Sigma) + 2mM Glutamine + 80 IU human Insulin/l (Novo) + 10% Foetal Bovine Serum (FBS) (heat-inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 34°C. Doubling time 48 hours. Cells show continuous growth at 34°C and growth arrest at 39°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service