Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ECV304

92091712, human bladder, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

ECV304, 92091712

biological source

human bladder

growth mode

Adherent

karyotype

High trisomic (modal no. 80)

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human urinary bladder carcinoma

Cell Line Description

Using new PCR based single locus fingerprint techniques, it was discovered that this cell line is identical to T-24. Because T-24 was established in 1970 compared to 1984 for ECV304 it is assumed that ECV304 is the false cell line. Originally, ECV304 was described by the originator as spontaneously transformed cell line derived from an umbilical cord from a new born Japanese female. Tumorigenic in Nude mice.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 9
D5S818: 10
D7S820: 10,11
THO1: 6
TPOX: 8,11
vWA: 17

Culture Medium

Medium 199 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA. 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service