NI 4 426/8B1-7
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity Anti-interleukin-4, human )
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 92030619-DNA-5UG
pkg of vial of cells 92030619-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
NS0
Reactivity
Anti-interleukin-4, human
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
Cell Line Origin
Mouse NS0 x Mouse BALB/c spleen
Cell Line Description
This neutralising antibody is specific for human IL-4 both natural and recombinant.
Immunogen
Recombinant human interleukin-4
Application
Immunoassays
Culture Medium
RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service