Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NI G 386/3D1-16

Reactivity: Anti-Granulocyte Colony Stimulating Factor (G-CSF), human, 92030613, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 92030613-DNA-5UG
pkg of vial of cells 92030613-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS0

Reactivity

Anti-granulocyte colony stimulating factor (G-CSF), human

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Not specified

Cell Line Description

This neutralising antibody is specific for human granulocyte colony stimulating factor (G-CSF), both recombinant and natural.

Immunogen

Human recombinant granulocyte colony stimulating factor (G-CSF)

Application

Immunoassays

Culture Medium

RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service