Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT3/B4

Reactivity: Anti-rh Interferon α-2 (recombinant human) and related subtypes, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 92030607

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 92030607-DNA-5UG
pkg of vial of cells 92030607-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8

Reactivity

Anti-rh interferon alpha-2 (recombinant human) and related subtypes

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Not specified

Cell Line Description

This monoclonal antibody showed strong neutralising activity against homologous IFN alpha-2. Cross-reactions with some h IFN alpha subtypes have been detected but no cross reaction with h IFN beta or gamma was shown.

Immunogen

Recombinant human interferon alpha-2

Application

Immuno assays, neutralisation of IFN alpha-2 activity.

Culture Medium

RPMI 1640 or DMEM + 2 mM glutamine + 7% Fetal Bovine Serum (FBS) (heat inactivated)

Subculture Routine

Maintain cultures between 1-5x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service