Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T11D7

Reactivity: Anti-Thy-1.1, 92021209

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

ImmunoDon

CBA mouse spleen

Myeloma

NS1

Reactivity

Anti-Thy-1.1

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x Mouse CBA spleen

Cell Line Description

The antibody is directed against the Thy-1.1 antigen of rats and mice. The antibody is cytotoxic and will cause lysis in a percentage of bone marrow, lymph node and spleen cells in vitro.

Immunogen

W/Fu rat thymocytes

Application

Depletion or isolation of T cell populations or rat B cell precursors

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service