J5
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 91072536, Reactivity: Anti-T Cell Activation Antigen, bovine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 91072536-DNA-5UG
pkg of vial of cells 91072536-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
P3X63Ag8
Reactivity
Anti-T Cell Activation Antigen, bovine
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse P3X63Ag8 x Mouse BALB/c spleen
Cell Line Description
The antibody is directed against a 100kDa antigen present on T lymphocytes activated by ConA, PWM, alloantigens or Theileria parva transformation.
Immunogen
Theileria parva infected T lymphoblast plasma membranes
Culture Medium
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells may require mouse macrophage feeder cells upon resuscitation.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service