Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IL-A43

Reactivity: Anti-CD2, bovine, 91072522, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 91072522-DNA-5UG
pkg of vial of cells 91072522-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-CD2, bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8.653 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against Bo CD2, the bovine equivalent of CD2. It shows species cross-reactivity with goat.

Immunogen

Bovine cytotoxic T cells

Application

Enumeration of bovine lymphocyte populations

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells may require mouse macrophage feeder cells upon resuscitation.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service