Key Documents
WES
91011612, chimpanzee skin, Fibroblast-like
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Product Name
WES, from chimpanzee skin, 91011612, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
biological source
chimpanzee skin
growth mode
Adherent
karyotype
Not specified
morphology
Fibroblast-like
receptors
Not specified
Cell Line Origin
Chimpanzee skin, SV40 transformed
Cell Line Description
A non-producer, Adeno 12-SV40 transformed chimpanzee skin cell line. The cell line has a high saturation density, exhibits colony formation in soft agar, is morphologically altered and produces tumours in nude mice.
Culture Medium
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:9, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.
Other Notes
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)