Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CCD 13Lu

90112704, human lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

CCD 13Lu, 90112704, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human lung

packaging

tube of 5 μg 90112704-DNA-5UG
pkg of vial of cells 90112704-1VL

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Storage temp.

−196°C

Cell Line Origin

Human lung

Cell Line Description

Established from lung tissue of a 71 year old male with pancreatic carcinoma and neoplastic invasion of the spleen and stomach and a mild case of emphysema. The cells can undergo up to 20 population doublings. Ethnicity: Black.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 12
D13S317: 10,14
D16S539: 9,11
D5S818: 10,12
D7S820: 10
THO1: 7,9.3
TPOX: 9,11
vWA: 14,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service