Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Jensen sarcoma

90111913, rat unknown/unspecified, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

Jensen sarcoma, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 90111913

biological source

rat unknown/unspecified

packaging

tube of 5 μg 90111913-DNA-5UG
pkg of vial of cells 90111913-1VL

growth mode

Adherent

karyotype

2n = 42, hyperdiploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat sarcoma, asparagine requiring

Cell Line Description

Used in studies relating to amino acid requirements and L-asparagine metabolism. Cloning studies show that variants not requiring L-asp are present in 100 per 1,000,000 cells Cloned populations which do not produce variants immediately do so after 6-8 further passages.

Application

Metabolic and cloning studies

Culture Medium

McCoy′s 5a + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Renew medium daily where possible. Subculture by placing culture in cold room (4°C) for 60 minutes, shake flask to suspend cells, centrifuge, resuspend in fresh medium and into fresh flasks. Split sub-confluent cultures (70-80%) 1:2 to 1:10; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service