Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

YN1/1.7.4

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 90110706, Reactivity: Anti-Murine Activated Lymphocyte Antigen-2 (MALA-2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Rat x Rat Hybridoma spleen

packaging

tube of 5 μg 90110706-DNA-5UG
pkg of vial of cells 90110706-1VL

ImmunoDon

F344 rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-Murine Activated Lymphocyte Antigen-2 (MALA-2)

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

Storage temp.

−196°C

Cell Line Origin

Rat Y3Ag1.2.3 x Rat F344 spleen

Cell Line Description

The antibody detects MALA-2 found primarily on activated T and B lymphocytes. The antigen has a molecular weight of 95-100kDa and is the murine equivalent of the human intercellular adhesion molecule ICAM-I. antibody directly inhibits proliferating cell populations in MLR.

Immunogen

NS1 cells

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service