Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LL86 (LeSa)

90102541, human lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

LL86 (LeSa), 90102541, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human lung

packaging

tube of 5 μg 90102541-DNA-5UG
pkg of vial of cells 90102541-1VL

growth mode

Adherent

karyotype

2n = 46; diploid

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian diploid lung

Cell Line Description

Derived from normal tissue of an 18 year old Caucasian male with osteogenic sarcoma.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 12
D13S317: 12
D16S539: 9,13
D5S818: 12,13
D7S820: 11
THO1: 6,7
TPOX: 8
vWA: 16,18

Culture Medium

Coon′s modified Ham′s F12 + Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. A further 10 population doublings expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service