Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Citrullinemia

90102532, human skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

Citrullinemia, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 90102532

biological source

human skin

packaging

tube of 5 μg 90102532-DNA-5UG
pkg of vial of cells 90102532-1VL

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian skin

Cell Line Description

Human diploid cell line derived from a young Caucasian female with Citrullinemia. The cells grow poorly when citrulline is substituted for arginine in the recommended medium and they incorporate very little 14C citrulline into cell protein. The cell line also has an abnormally low affinity for citrulline as a substrate in an in vivo assay of arginosuccinate synthetase.

Application

Genetic studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 2% Non Essential Amino Acids (NEAA) + 2% Vitamins + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Finite lifespan up to 30 passages.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service