Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DH14

90070551, insect embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

DH14, 90070551, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

insect embryo

packaging

tube of 5 μg 90070551-DNA-5UG
pkg of vial of cells 90070551-1VL

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Drosophila embryo

Cell Line Description

Derived from Drosophila embryo, can support the growth of picornaviruses and rhabdoviruses.

Application

Virus studies

Culture Medium

Schneider′s Insect Medium + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. Cells may semi-adhere to tissue culture flask, shake gently to dislodge; 25°C; No CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service