Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CCD-18Co

90070503, human colon, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

Product Name

CCD-18Co, 90070503

biological source

human colon

growth mode

Adherent

karyotype

modal No 46

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon fibroblast, normal, ethnicity: Black

Cell Line Description

Derived from biopsy colon tissue of a black female infant (2.5 months old). CCD 18Lu (90112706) and CCD-18Co (90070503) were derived from the lung and colon respectively from the same patient; this has been confirmed by STR profiling at ECACC.

Application

Aging studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 8
D13S317: 12
D16S539: 12,13
D5S818: 12
D7S820: 8
THO1: 6,7
TPOX: 8,11
vWA: 15,17

Culture Medium

EMEM (EBSS) + 2mM glutamine + 1% NEAA + 10% FBS

Subculture Routine

Split confluent cultures 1:2 to 1:3 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service