Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Vx2

90030912, rabbit unknown/unspecified, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

Product Name

Vx2, 90030912

biological source

rabbit unknown/unspecified

growth mode

Adherent

morphology

Fibroblast-like

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see Vx2

Cell Line Origin

Rabbit tumour

Cell Line Description

From a rabbit Orycytolagus cuniculus tumour of unspecified tissue origin.

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 split ratio i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service