Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

161BR

90011810, human skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

161BR, 90011810

biological source

human skin

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human skin fibroblast

Cell Line Description

Established from a skin of a normal 49 year old male. The cells have been used in radiation survival studies.

Application

DNA repair studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 12,14
D16S539: 11
D5S818: 9,11
D7S820: 8,13
THO1: 6,9.3
TPOX: 8
vWA: 17,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service