Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D17

89090403

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see D17

Cell Line Origin

Canine Poodle primary osteosarcoma

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service