Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U373 MG Cell Line human

89081403, human brain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

U373 MG Cell Line human, 89081403, Isolated from brain epithelium

biological source

human brain

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Derived from a malignant tumor by explant technique.
Subculture: Spit at 70-80% confluency, approx. 1:3 to 1:6, seed at 2-4 × 104 viable cells/cm2. Trypsinize using 0.25% solution, with or without EDTA; 37 °C and 5% CO2.
Recommended culture medium: EMEM (EBSS) + 2 mM L-Glutamine + 1% Non-essential amino acids (NEAA) + 1 mM Sodium pyruvate + 10% FBS.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service