Key Documents
EB176 (JC)
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
biological source
chimpanzee blood
packaging
tube of 5 μg 89072704-DNA-5UG
pkg of vial of cells 89072704-1VL
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblast
receptors
Not specified
Cell Line Origin
Chimpanzee peripheral lymphoblast
Cell Line Description
Epstein Bar virus (EBV) transformed lymphoblastoid cell line from the peripheral lymphocytes of a male Chimpanzee.
Culture Medium
RPMI 1640 (R0883)+ 2 mM L-Glutamine (G7513) + 15% FBS / FCS (F2442).
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; do not over dilute; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)