Key Documents
EB (JC)
89072703, gorilla blood, Lymphoblast
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Product Name
EB (JC), from gorilla blood, 89072703, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
biological source
gorilla blood
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblast
receptors
Not specified
Cell Line Origin
Gorilla peripheral lymphoblast
Cell Line Description
Epstein Bar virus (EBV) transformed lymphoblastoid cell line from the peripheral lymphocytes of a female Gorilla.
Culture Medium
RPMI 1640 (R0883)+ 2 mM L-Glutamine (G7513) + 15% FBS / FCS (F2442).
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; do not over dilute; 5% CO2; 37 °C. Cells show typical lymphoblast morphology, i.e., they grow in large spheroid clumps as well as single cells in suspension.
Other Notes
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)