Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ICR-2A

89072615, frog embryo, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

ICR-2A, 89072615, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

frog embryo

packaging

tube of 5 μg 89072615-DNA-5UG
pkg of vial of cells 89072615-1VL

growth mode

Adherent

karyotype

2n = 26, Stable haploid

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Frog Grass embryo

Cell Line Description

Previously designated RPH68.2A. Derived from androgenetic haploid embryos of the grass or leopard frog, (Rana pipiens). The cells retain their haploid chromosome complement for over 200 generations. Used in genetic studies.

Application

Genetic studies

Culture Medium

L15 (50%) + Distilled Water (40%) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 25°C. Do not use CO2.Grow cells in dark (i.e wrapped in foil). Cells do not reach 100% confluency, only approximately 70-80%. Freeze in glycerol.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service