Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Mc 39-16

Reactivity: Anti-A31 idiotype, murine, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 89072609

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Rat Heterohybridoma

packaging

tube of 5 μg 89072609-DNA-5UG
pkg of vial of cells 89072609-1VL

ImmunoDon

LOU rat spleen

Myeloma

NS1

Reactivity

Anti-A31 idiotype, murine

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x Rat LOU spleen

Cell Line Description

This heterohybridoma secretes a rat monoclonal antibody against the A-31 idiotype of murine B cell lymphoma. Available for use as a heterohybridoma secreting rat IgG2a antibody. However, use in studies of the A31 tumour requires permission from the originators.

Immunogen

Sheep anti murine IgM-A31 IgM complexes

Application

The isotype of the monoclonal antibody produced by this cell line is rat IgG2a.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service