Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TE 671

89071904, human embryo, Spindle shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

Product Name

TE 671, 89071904

biological source

human embryo

growth mode

Adherent

morphology

Spindle shaped

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see TE 671

Cell Line Origin

Human Caucasian rhabdomyosarcoma

Cell Line Description

TE671 is now known to be identical to the human rhabdomyosarcoma RD cell line (Sigma Catalogue number. 85111502). Although originally described as derived from a cerebellar medulloblastoma, the cell line TE 671 is now known to be identical to the human rhabdomyosarcoma RD cell line (Sigma Catalogue number. 85111502). TE671 should be considered to be derived from RD.

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service