Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

YTS 156.7.7

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-Lyt-3 epitope B, mouse, 89040604

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Rat x Rat Hybridoma

packaging

tube of 5 μg 89040604-DNA-5UG
pkg of vial of cells 89040604-1VL

ImmunoDon

DA rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-Lyt-3 epitope B, mouse

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

Storage temp.

−196°C

Cell Line Description

The antibody is directed against the B epitope of the mouse Lyt-3 (CD8) antigen.Has been used in CD8 subset depletion in vivo and flow cytometry studies. Cambridge Enterprise Reference number AB15: www.enterprise.cam.ac.uk.

Immunogen

CBA mouse thymocytes

Culture Medium

DMEM + 2 mM glutamine + 0.05 mM 2-mercaptoethanol (2ME) + 20% Fetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37 °C. Saturation density 9 x 105/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service