YBM 15.1.6
89040602, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-MAC-1, mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Rat x Rat Hybridoma
packaging
tube of 5 μg 89040602-DNA-5UG
pkg of vial of cells 89040602-1VL
ImmunoDon
DA rat spleen
Myeloma
Y3Ag1.2.3
Reactivity
Anti-MAC-1, mouse
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat Y3Ag1.2.3 x Rat DA spleen
Cell Line Description
The antibody is directed against the mouse MAC-1 antigen, the murine equivalent of CD11b.
Immunogen
CBA mouse bone marrow
Culture Medium
IDMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells may adhere to the plastic flask - may require trypsin or shaking to remove them
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service