Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

hamster ovary

packaging

tube of 5 μg 89030316-DNA-5UG
pkg of vial of cells 89030316-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Ornithine decarboxylase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese ovary

Cell Line Description

Overproduces ornithine decarboxylase in response to polyamine deprivation.

Application

Enzymatic studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + Ham′s F12 (without Putrescine) (1:1) + 0.1% BSA.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 5x1,000 to 1x10,000 cells/cm2 using cold PBS and 0.25% cold trypsin. Place flasks at 4°C for 5-10 minutes or until cells detach. Culture cells at 37°C; 5% CO2. Pre-soak flasks in porcine

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service